Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LONRF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA535914
Description
Recommended positive controls: U87-MG, SK-N-SH, IMR-32, SK-N-AS. Predicted reactivity: Human (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
LONRF2 contains one Lon domain, one RING type zinc finger and six TPR repeats. The function of LONRF2 is unknown. There are two named isoforms.
Specifications
| LONRF2 | |
| Polyclonal | |
| Unconjugated | |
| LONRF2 | |
| LON peptidase N-terminal domain and ring finger 2; LON peptidase N-terminal domain and RING finger protein 2; LONRF2; Neuroblastoma apoptosis-related protease; RING finger protein 192; RNF192 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 164832 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q1L5Z9 | |
| LONRF2 | |
| Recombinant protein encompassing a sequence within the center region of human LONRF2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction