Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LONRF3 Monoclonal Antibody (1D5)
GREENER_CHOICE

Catalog No. PIMA522606
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522606 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522606 Supplier Invitrogen™ Supplier No. MA522606
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MESVRIEQML SLPAEVSSDN LESAERGASA AQVDMGPHPK VAAEGPAPLP TREPEQEQSP GTSTPESKVL LTQADALASR GRIREALEVY.

The protein encoded by this gene contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. Multiple alternatively spliced transcript variants have been suggested, but their full length natures are not clear.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LONRF3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1D5
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene LONRF3
Gene Accession No. Q496Y0
Gene Alias LON peptidase N-terminal domain and ring finger 3; LON peptidase N-terminal domain and RING finger protein 3; LONRF3; RING finger protein 127; RNF127
Gene Symbols LONRF3
Host Species Mouse
Immunogen LONRF3 (NP_079054, 1 a.a. ~ 90 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79836
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.