Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LONRF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584482
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584482 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584482 Supplier Invitrogen™ Supplier No. PA584482
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MESVRIEQML SLPAEVSSDN LESAERGASA AQVDMGPHPK VAAEGPAPLP TREPEQEQSP GTSTPESKVL LTQ Highest antigen sequence indentity to the following orthologs: Mouse - 47%, Rat - 47%.

The protein encoded by this gene contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. Multiple alternatively spliced transcript variants have been suggested, but their full length natures are not clear.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LONRF3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LONRF3
Gene Accession No. Q496Y0
Gene Alias LON peptidase N-terminal domain and ring finger 3; LON peptidase N-terminal domain and RING finger protein 3; LONRF3; RING finger protein 127; RNF127
Gene Symbols LONRF3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LONRF3. Recombinant protein control fragment (Product #RP-108451).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79836
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.