Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ AccuGENE™ 1M Tris HCl Buffers

Catalog No. bma51237
Click to view available options
pH:
7.2
7.4
8.0

Catalog No. BMA51237

Supplier: Lonza™ 51237

Only null left
Add to Cart
Add to Cart

Ready-to-use solutions ideal for a wide range of molecular biology applications

  • Prepared with 18 megohm water
  • Filtered using 0.2μ filter and filled into sterile bottles
  • Manufactured according to strict quality control standards to ensure consistency
  • Guaranteed DNase, RNase and protease-free
  • Customized solutions are available to meet individual needs

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 M
DNase Yes
For Use With (Application) Ideal for a wide range of molecular biology applications
pH 7.4
Quantity 1000 mL

For research or manufacturing use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.