Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ BioWhittaker™ HEPES Buffer

Catalog No. p-4892982
Click to view available options
Quantity:
100 mL
500 mL

Used in open vessels incubated under atmospheric conditions. BioWhittaker™ HEPES Buffer is manufactured in accordance with cGMP regulations.

 

  • Used in open vessels incubated under atmospheric conditions
  • Behaves as zwitterion
  • Sodium chloride concentration of HEPES buffered medium has been reduced to maintain same Osmolality of original media formulations
  • Manufactured in accordance with cGMP regulations
  • Cell culture-tested to assure compatibility

 

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15° to 30°C
Product Type HEPES Buffer
Sterility Sterile
pH 6.98 to 7.3
Quantity 500 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.