Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lonza™ BioWhittaker Insect-XPRESS Insect Cell Culture Media (1X) with L-Glutamine
Click to view available options
Quantity:
1 L
With Additives:
Glutamine
Description
- Protein-free
- Cell densities in excess of 8.3×106 cells/mL can be achieved with suspension cultures of Sf9 cells using Insect-XPRESS Medium and excess of oxygen
- Formulation can also be used for stationary mono¬layer cultures and shake-flask cultures
Compliance
Manufactured in accordance with cGMP regulations for insect cell lines; formulations recommended by the Tissue Culture Association.

Specifications
Specifications
Form | Liquid |
Product Type | Insect Cell Medium, Protein-free |
With Additives | Glutamine |
Quantity | 1 L |
Content And Storage | 2° to 8°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction