Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ BioWhittaker™ Insect Cell Culture Media: Schneider's Drosophila Medium (Modified)

Catalog No. bw04351q
Click to view available options
Quantity:
1 L

formulated for the growth of S2 cells from the fruit fly, Drosophila melanogaser. BioWhittaker™ Insect Cell Culture Media: Schneider's Drosophila Medium (Modified) may be used to culture a variety of other insect cell lines as well.

Used for:
  • Drosophila melanogaser
  • Other insect cell lines
  • Transient or stable transfection
  • Large scale protein production in S2 cells
Contains:
  • malic acid
  • fumaric acid
  • alpha-ketoglutaric acid
  • succinic acid
  • sugar trehelose
  • Manufactured in accordance with cGMP regulations
  • The formulations used for standard liquid media products are those recommended by the Tissue Culture Association

Compliance

Manufactured in accordance with cGMP regulations for insect cell lines; formulations recommended by the Tissue Culture Association.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 L
Content And Storage 2° to 8°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.