Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BioWhittaker™ Water for Cell Culture, Lonza™

Catalog No. p-2894722
Click to view available options
Quantity:
500 mL
1 L

Water for Injection (WFI) system for culture applications. BioWhittaker™ Water for Cell Culture is manufactured aseptically under cGMP conditions using a validated sterile filtration method.

  • Method for the production of WFI includes:
  • Ultra filtration
  • Reverse osmosis
  • Mixed bed deionizers
  • Hot recirculating loop and cooled at point of use
  • System is validated and monitored for bioburden, endotoxin, total organic carbons and conductivity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Quantity 500 mL
Chemical Name or Material Water
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.