Learn More
Lonza™ FlashGel™ System DNA Markers
Description
- Ready-to-load molecular weight standards
- Separates DNA in real time: 2 to 7 minutes
- Clean, sharp separation and straight, uniform sample lanes
- Watch DNA migration as it happens: Monitor gel runs right at the bench, without UV light
- Highly sensitive system allows a reduction in DNA levels, saving time and money
- Outstanding resolution, exquisite sensitivity: 5 to 20 times more sensitive than EtBr
- Detect <0.1ng DNA or <10ng total RNA
- Recommended for FlashGel DNA cassettes
FlashGel 1.2% DNA Marker
- Sizes: 100/200/300/500/800/1250/2000/4000bp
- Recommended for 1.2% FlashGel DNA cassettes
- Sizes: 50/100/150/200/300/500/800/1500bp
- Recommended for 2.2% FlashGel DNA cassettes
- Sizes: 100/300/500/800/1500/3000bp
- Recommended for double tier FlashGel DNA cassettes
- For research or manufacturing use only
- Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
Specifications
Specifications
| Form | Liquid |
| For Use With (Application) | Double-tier FlashGel DNA Cassette |
| No. of Bands | 6 |
| Quantity | 500 μL |
| Storage Requirements | 4°C |
| Product Type | DNA Molecular Weight Marker/Ladder |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.