Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lonza™ FlashGel™ System DNA Markers
Click to view available options
No. of Bands:
6
8
For Use With (Application):
1.2% FlashGel DNA Cassette
2.2% FlashGel DNA Cassette
Double-tier FlashGel DNA Cassette
Description
- Ready-to-load molecular weight standards
- Separates DNA in real time: 2 to 7 minutes
- Clean, sharp separation and straight, uniform sample lanes
- Watch DNA migration as it happens: Monitor gel runs right at the bench, without UV light
- Highly sensitive system allows a reduction in DNA levels, saving time and money
- Outstanding resolution, exquisite sensitivity: 5 to 20 times more sensitive than EtBr
- Detect <0.1ng DNA or <10ng total RNA
- Recommended for FlashGel DNA cassettes
FlashGel 1.2% DNA Marker
- Sizes: 100/200/300/500/800/1250/2000/4000bp
- Recommended for 1.2% FlashGel DNA cassettes
- Sizes: 50/100/150/200/300/500/800/1500bp
- Recommended for 2.2% FlashGel DNA cassettes
- Sizes: 100/300/500/800/1500/3000bp
- Recommended for double tier FlashGel DNA cassettes
- For research or manufacturing use only
- Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals

Specifications
Specifications
Product Type | DNA Molecular Weight Marker/Ladder |
Form | Liquid |
Formulation | 5μL Load 1:5 dilution |
For Use With (Application) | 1.2% FlashGel DNA Cassette |
No. of Bands | 8 |
Quantity | 500 μL |
Storage Requirements | 4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction