Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ Latitude™ Precast Agarose Midigels

Catalog No. p-121038
Click to view available options
Separation Range:
100 to ≥2000 bp
300 to ≥8000 bp
Wells:
2 x 20-well
20-well

Efficiently resolve DNA ranging from 10bp to more than 10kb. Latitude™ Precast Agarose Midigels are designed for accurate DNA fragment separation for most any gel electrophoresis application.

  • Ideal for checking restriction digests and PCR products
  • Quality and performance tested to ensure reproducible results every time
  • Prestained with ethidium bromide
  • Compatible with common midichambers, multichannel pipets and alternate-well microtiter plate spacing

Note:ethidium bromide is a powerful mutagen; gloves should be worn when opening pouches and handling gels and solutions containing ethidium bromide

TRUSTED_SUSTAINABILITY

Specifications

Buffer TBE
Color Clear
Concentration 1%
Product Type Precast Gel
Separation Range 300 to ≥8000 bp
Wells 2 x 20-well
Dnase Free Yes
Formulation 1% SeaKem™ LE Plus Agarose, 1X TBE, prestained with 0.5μg/mL of Ethidium Bromide
For Use With (Application) DNA separation by gel electrophoresis
Material Agarose
Quantity 8 Gels
RNase Free Yes
Storage Requirements 18° to 26°C
Thickness (English) 0.23 in.
Thickness (Metric) 0.6 cm
Volume Per Well 10 to 12 μL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.