Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ Reliant™ Precast RNA Gels

Catalog No. p-310891
Click to view available options
Wells:
20-well
8-well

Formulated for separation of RNA of sizes from 0.25 to at least 10kb. Reliant™ Precast RNA Gels are ready-to-use RNase free agarose gels suitable for a variety of RNA applications including Northern blotting and analysis of RNA transcripts.

  • Ready to use
  • RNase free
  • Free of denaturants, eliminating problems associated with staining gels containing formaldehyde
  • Suitable for a variety of applications, including Northern blotting and analysis of RNA transcripts
  • Compatible with most common mini chambers
  • Available with a separate container of glyoxal buffer
TRUSTED_SUSTAINABILITY

Specifications

Buffer MOPS
Concentration 1.25%
Format Multiwell
Product Type Precast Gel
Wells 8-well
Dnase Free Yes
Formulation 1.25% SeaKem™ Gold Agarose, 1X MOPS
For Use With (Application) Northern blotting and analysis of RNA transcripts
Material Agarose
Quantity 20 Gels
RNase Free Yes
Storage Requirements 18° to 26°C
Thickness (English) <0.19 in.
Thickness (Metric) <0.5 cm
Volume Per Well <15 μL
Length (English) Gel 3.7 in.
Length (Metric) Gel 9.5 cm
Width (English) Gel 2.37 in.
Width (Metric) Gel 6 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.