Learn More
SeaKem™ GTG™ Agarose, Lonza™
Catalog No. p-2861586
Click to view available options
Quantity:
25 g
125 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy
Specifically designed for preparative DNA electrophoresis. Lonza™ SeaKem™ GTG™ Agarose is a standard gelling temperature, high gel strength agarose that resolves DNA fragments greater than 1,000 bp.
  • DNA recovered from SeaKem™ GTG™ Agarose gels may be reliably digested and ligated
  • Extensively performance tested to ensure complete compatibility with routine molecular biology techniques
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material SeaKem GTG Agarose
Quantity 25 g
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.