Learn More
Hardy Diagnostics™ Disposable Inoculating Loop
Shop All Hardy Diagnostics Products
Click to view available options
Volume:
1 μL
Description
- Sterile
- Free of lubricants, oils, and electrostatic charges
- Available in choice of flexible or rigid plastic
- Color coded for easy product identification
- Packed in resealable plastic pouches
Specifications
Specifications
| Disposable | Single-use |
| Volume | 1 μL |
| Sterility | Sterile |
| Diameter (English) Inside | 1 μL |
| Material | Plastic |
| Product Type | Inoculating Loop |
| Quantity | 40/Pk., 1000/Box |