Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LOR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542059
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542059 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542059 Supplier Invitrogen™ Supplier No. PA542059
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GYSGGGCGGG SSGGGGGGGI GGCGGGSGGS VKYSGGGGSS GGGSGCFSSG Sequence homology: Cow: 93%; Guinea Pig: 75%; Human: 100%; Mouse: 86%.

This gene encodes loricrin, a major protein component of the cornified cell envelope found in terminally differentiated epidermal cells. Mutations in this gene are associated with Vohwinkel's syndrome and progressive symmetric erythrokeratoderma, both inherited skin diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LOR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LORICRIN
Gene Accession No. P23490
Gene Alias AI036317; LOR; Loricrin; loricrin cornified envelope precursor protein; LRN; MGC111513; RGD1559993; S77319
Gene Symbols LORICRIN
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human LOR (aa 58-107).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4014
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.