Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Loratadine, 98+%

Catalog No. aaj60190me
Click to view available options
Quantity:
500 mg
1 g
5 g

A nonsedating-type histamine H1-receptor antagonist | CAS: 79794-75-5 | C22H23ClN2O2 | 382.888 g/mol

Loratadine, is used as a peripheral histamine H1 receptor agonist. It is also used as an orally active antiallergic agent. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Loratadine, is used as a peripheral histamine H1 receptor agonist. It is also used as an orally active antiallergic agent. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
It is soluble in DMSO (50 mg/ml), ethanol (77 mg/ml at 25°C), water (<1mg/mL at 25°C), chloroform, and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away fom strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79794-75-5
Molecular Formula C22H23ClN2O2
Molecular Weight (g/mol) 382.888
MDL Number MFCD00672869
InChI Key JCCNYMKQOSZNPW-UHFFFAOYSA-N
Synonym loratadine, claritin, loratidine, alavert, clarityn, lisino, clarityne, loracert, loradex, bonalerg
PubChem CID 3957
IUPAC Name ethyl 4-(8-chloro-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-ylidene)piperidine-1-carboxylate
SMILES CCOC(=O)N1CCC(=C2C3=C(CCC4=C2N=CC=C4)C=C(C=C3)Cl)CC1

Specifications

Melting Point 134°C
Color White
Quantity 500 mg
Merck Index 14,5578
Solubility Information It is soluble in DMSO (50mg/ml),ethanol (77mg/ml at 25°C),water (<1mg/ml at 25°C),chloroform,and methanol.
Formula Weight 382.88
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Loratadine
RTECSNumber TM6129200
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.