Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Losartan potassium salt

Catalog No. p-7139371
Click to view available options
Quantity:
5 g
25 g

Catalog No. AC462740050

Supplier: Thermo Scientific Chemicals 462740050

CAS: 124750-99-8 | C22H22ClKN6O | 461.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124750-99-8
Molecular Formula C22H22ClKN6O
Molecular Weight (g/mol) 461.01
InChI Key OXCMYAYHXIHQOA-UHFFFAOYSA-N
PubChem CID 53396294

Specifications

Quantity 5 g
Chemical Name or Material Losartan potassium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.