Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LOTUS Mouse anti-Human,Mouse,Rat, Unconjugated, Clone: 52-1E, ITM, MilliporeSigma™

Catalog No. MABN25491MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN25491MI 100 μg
MABN25492MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN25491MI Supplier MilliporeSigma Supplier No. MABN2549100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LOTUS Monoclonal antibody specifically detects LOTUS in Human,Mouse,Rat samples. It is validated for Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LOTUS
Applications Western Blot
Classification Monoclonal
Clone 52-1E, ITM
Conjugate Unconjugated
Gene Alias Lateral olfactory tract usher substance;Cartilage Acidic Protein–1B
Host Species Mouse
Immunogen A recombinant fragment of LOTUS protein consisting of several specific amino acids from the N-terminal region of rat lateral olfactory tract usher substance (LOTUS).
Molecular Weight of Antigen ∼75 kDa observed. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 100 μg
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone 52-1E, ITM is a mouse monoclonal antibody that detects Lateral olfactory tract usher substance (LOTUS) protein. it targets an epitope within the N-terminal region.
Target Species Human, Mouse, Rat
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.