Learn More
Lovibond™ Aqua Comparitor Color Discs: Chlorine
Catalog No. p-4909448 Shop All Tintometer Group Products
Click to view available options
Range:
0 to 1mg/L
0 to 3.5mg/L
0 to 4mg/L
0.01 to 0.2mg/L
0.02 to 0.3 mg/L
0.1 to 2mg/L
10 to 300mg/L
Type:
Chlorine Color Disc
Chlorine Dioxide Color Disc
For testing water with the Aqua Comparator
Economical replacement discs make it easy to test for chlorine using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
For Use With (Application) Testing water with the Aqua comparator™
For Use With (Equipment) Aqua comparator™
Range 0.02 to 0.3 mg/L
Type Chlorine Color Disc
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.