Learn More
Lovibond™ Aqua Comparitor Color Discs: Copper
Shop All Tintometer Group Products
Click to view available options
Range:
0 to 1mg/L
0 to 5mg/L
0.01 to 0.2mg/L
Type:
Copper HR Color Disc
Copper HR Powder Color Disc
Copper LR Color Disc
Copper LR Powder Color Disc
Description
For testing water with the Aqua Comparator
Economical replacement discs make it easy to test for copper using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Specifications
| For Use With (Application) | Testing water with the Aqua comparator™ |
| For Use With (Equipment) | Aqua comparator™ |
| Range | 0 to 5mg/L |
| Type | Copper HR Color Disc |