Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ CHECKIT™ Comparator Test Kits: Chlorine-Free

Catalog No. 09053108 Shop All Tintometer Group Products
Click to view available options
Range:
0 to 3.5mg/L

Includes

  • Aqua Comparator Housing
  • Aqua Comparator Disc
  • DPD Free Reagent, 5ml sample (100 pack)
  • 2 x 13.5mm Sample Cells
  • Tablet Crusher
  • Instructions
  • Carrying Case

Accurate test kits for routine water analysis

  • Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value

Specifications

Accuracy ±5%
For Use With (Application) Routine water analysis
Includes Aqua comparator™ housing, aqua comparator™ disc, DPD free reagent, 5mL sample, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case
No. of Tests 100
Range 0 to 3.5mg/L
Type Chlorine Free and Total Test Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.