Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Liquid Reagents for Water Analysis

Catalog No. p-4684679 Shop All Tintometer Group Products
Click to view available options
Color:
Colorless
Dark Orange
pH Range:
8
8.6
13.7
Volume (English):
1.01 oz.
16.9 oz.
Volume (Metric):
30 mL
500 mL

Specifications

Color Dark Orange
pH Range 8
Volume (Metric) 500 mL
Format Liquid
For Use With (Application) Water analysis
For Use With (Equipment) SC450 manganese, MC500 colorimeter, SP600 spectrophotometer, manganese reagent set
Range 0 to 0.7mg/L
Storage Requirements 20°C
Type PAN Indicator Solution
Volume (English) 16.9 oz.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.