Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ MD 100 Boiler Water Colorimeter

Catalog No. 09053675
Click to view available options
Resolution:
0.01 A

Includes

Instrument in carrying case, 4 x AA batteries, three round glass vials (24mm) with lids, crushing/stirring rod, Guarantee sheet, Certificate (COC), Instruction manual

One colorimeter tests multiple parameters associated with boiler water.

One photometer tests multiple parameters associated with boiler water.

  • Ideal for routine, on-site system monitoring
  • Colorimetric measurement for accurate, repeatable results
  • Easy-to-use, push button technology with step by step guides
  • Negates the need for up to 13 seperate pieces of equipment
  • Select range of required reagents to complete your kit
  • Combine with pH, conductivity and drop test chemistries - complete analysis program
  • Supplied pre-calibrated with test methods

Specifications

  • Optics: LEDs, interference filters (IF) and photo sensor in transparent sample chamber.
  • Wavelength Accuracy: ±1 nm
  • Photometric Accuracy: 3% FS (T = 2° to 25°C)
  • Power Supply: 4 micro batteries (AAA), capacity approx. 17 hours or 5000 tests
  • Auto - OFF: automatic switch-off
  • Display: Backlit LCD (on keypress)
  • Storage: internal ring memory for 16 data sets
  • Interfaces: Infrared interface for test data transfer
  • Additional feature: real time clock and date
  • Calibration: factory calibration and user calibration
  • Dimensions (L x W x H): 155 x 75 x 35 mm
  • Weight: basic unit approx. 260 g

Certifications

CE marked

Specifications

Accuracy 3% Full scale
Calibration Yes
Certifications/Compliance CE
Data Storage Internal ring memory for 16 data sets
Display Backlit LCD
Dimensions (L x W x H) 155 x 75 x 35 mm
Light Source LED
For Use With (Application) Testing multiple parameters associated with boiler water
Height (English) 1.3 in.
Height (Metric) 35 mm
Includes Instrument in carrying case, 4 x AA batteries, three round, 24mm dia. glass vials with lids, crushing/stirring rod, Guarantee sheet, Certificate (COC), Instruction manual
Length (English) 6.1 in.
Length (Metric) 155 mm
Resolution 0.01 A
Temperature Range (Metric) 5°C to 40°C
Weight (English) 0.5 lb.
Weight (Metric) 260 g
Width (English) 3 in.
Width (Metric) 75 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.