Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lovibond™ Powder Packs™ for Free Chlorine
Shop All Tintometer Group Products
Click to view available options
Quantity:
100 Pack
1000 Pack
Range:
0 to 2 mg/L
0 to 3.5 mg/L
0 to 5 mg/L
Volume (English):
0.16 oz.
0.33 oz.
0.84 oz.
Volume (Metric):
5 mL
10 mL
25 mL
Description
Premeasured unit dose DPD reagents for water quality testing
- For use in SC450™ Colorimeters and Aqua Comparator™ Test Kits
- Also suitable for use in Hach™ instruments and test kits
- Easy-to-open, sealed foil packets eliminate risk of cross-contamination
- Available for 5, 10, and 25mL sample sizes
- Extended shelf life of up to 5 years
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Compliance
US EPA-accepted DPD method
Specifications
Specifications
Certifications/Compliance | US EPA |
Volume (Metric) | 10 mL |
Format | Powder |
For Use With (Application) | Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing |
For Use With (Equipment) | SC450 colorimeter and Aqua Comparator™ test kits, Hach™ instruments and test kits, spectrophotometer |
Quantity | 100 Pack |
Range | 0 to 2 mg/L |
Type | DPD Free Chlorine Reagent |
Volume (English) | 0.33 oz. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction