Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Powder Packs™: Silica HR Citric Acid

Catalog No. p-4641523
Click to view available options
Quantity:
100 Pack
Range:
0 to 1.6mg/L
Volume (Metric):
10 mL

Pre-measured, unit dose reagents for water quality testing

  • Simply tear open Powder Pack™ and pour reagent into sample
  • Large design makes them easier to handle
  • “Easy Open” notch makes it simple to tear open the package
  • Easily create a “V”, to allow user to pour reagent into sample
  • Beveled edge prevents reagent from getting stuck in the corners

Specifications

Color White
Composition Citric Acid
pH Range 2.2
Volume (Metric) 10 mL
Format Powder
For Use With (Application) Water quality testing
For Use With (Equipment) Hach™ and other manufacturers test kits, colorimeters and spectrophotometers
Quantity 100 Pack
Range 0 to 1.6mg/L
Storage Requirements 20°C
Type Silica HR Citric Acid Reagent
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.