Learn More
Lovibond™ Sample Cells for SP600 Spectrophotometer
Catalog No. p-4779854 Shop All Tintometer Group Products
Click to view available options
Type:
Sample Cell without Lid
Pathlength:
10 mm
50 mm
Replacement sample cell for SP600 spectrophotometer.
  • Made from optical glass
Specifications
Type Sample Cell without Lid
For Use With (Equipment) SP600 spectrophotometer
Pathlength 50 mm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.