Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Solution: PAN; 0.1%

Catalog No. p-4784910 Shop All Tintometer Group Products
Click to view available options
Quantity:
50 mL
100 mL

Pre-measured, unit dose reagent for water quaility testing

  • Specifically formulated to serve as direct replacement to Hach™ PAN Indicator Solution

Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing

Specifications

Certifications/Compliance US EPA
Composition N,N-dimethylformamide 40 to 50%, Octylphenol Decaethylene 5 to 10%, distilled Water, conductivity or of similar purity 20 to 35%, non-hazardous components >5%
Volume (Metric) 50 mL
For Use With (Application) Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Quantity 50 mL
Range 0.01 to 0.7mg/L
Test Method PAN
Type PAN Indicator Solution
Volume (English) 1.69 oz.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.