Learn More
IBI Scientific™ Low Melting Point Agarose
Catalog No. p-4911316
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
500 g
Designed for preparative DNA and RNA electrophoresis, cloning of tissue culture cells and viral plague assays, and fragment recovery following electrophoresis. IBI Scientific™ Low Melting Point Agarose has lower gelling point and melting point when compared to Standard Agarose allows quick and thorough digestion during extraction procedures and easy in-gel manipulations.
  • Lower gelling and melting temperature than standard agarose
  • Lower gel strength than standard agarose while still easy to handle
  • Higher gel transparency than standard agarose
  • Greater sieving capacity than standard agarose
  • Separates and finely resolves molecules >1000bp
  • DNase- and RNase-Free
Specifications
Product Type Agarose
For Use With (Application) Used as gel matrix and preparation of large DNA samples for plugs
Quantity 100 g
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.