Learn More
MilliporeSigma™ Supelco™ Low Volume Static Mixer (50 to 250 μL) Housing, Binary
$684.00 - $790.00
Specifications
Description Fitting: 10-32 ASI in-line housing for 1/16 in. O.D. tubing
For Use With (Equipment) ASI Low Volume Mixer Cartridge
Volume (Metric) 50 to 250 μL
View More Specs
Products 2
Catalog Number Mfr. No. Material Price Quantity & Availability  
Catalog Number Mfr. No. Material Price Quantity & Availability  
11-100-3599
View Documents
MilliporeSigma™ Supelco™
500518
PEEK
Each for $790.00
Only null left
Add to Cart
11-100-3701
View Documents
MilliporeSigma™ Supelco™
57549
Stainless Steel
Each for $684.00
Only null left
Add to Cart
Description
Description
Specifications
Specifications
Fitting: 10-32 ASI in-line housing for 1/16 in. O.D. tubing
ASI Low Volume Mixer Cartridge
50 to 250 μL
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.