Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Lowenstein-Jensen Medium (LJ)
Click to view available options
Quantity:
100/Pk.
20/Pk.
Format:
1 oz. Bottle
20mm x 150mm Tube - Slant
Tube
Media Type:
Lowenstein-Jensen Medium (LJ)
Manufacturing Origin:
United States
Description
Cultivate and isolate Mycobacterium spp. using Thermo Scientific™ Lowenstein-Jensen Medium (LJ).
Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use tubes and bottles of Lowenstein-Jensen Medium, a glycerated egg-potato medium which provides fatty acids and protein required for metabolsm of mycobacteria. Malachite green is added to inhibit other mycobacteria and as a pH indicator.
Thermo Scientific Lowenstein-Jensen Medium (LJ) agar slants are available in a variety of fill volumes and pack sizes.
Mycobacteria most commonly involve infections of the pulmonary system, but some species infect the lymphatic system and skin. Mycobacteria are gram-positive, nonmotile, acid-fast pleomorphic rods. They vary in colony morphology, pigmentation, and growth rates. Mycobacterium tuberculosis, the most important pathogenic Mycobacterium, is slow-growing and causes highly contageous, airborne respiratory disease. Tuberculosis is responsible for an estimated 2 million deaths per year worldwide.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Thermo Scientific Lowenstein-Jensen Medium (LJ) agar slants are available in a variety of fill volumes and pack sizes.
Mycobacteria most commonly involve infections of the pulmonary system, but some species infect the lymphatic system and skin. Mycobacteria are gram-positive, nonmotile, acid-fast pleomorphic rods. They vary in colony morphology, pigmentation, and growth rates. Mycobacterium tuberculosis, the most important pathogenic Mycobacterium, is slow-growing and causes highly contageous, airborne respiratory disease. Tuberculosis is responsible for an estimated 2 million deaths per year worldwide.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Format | 1 oz. Bottle |
| Manufacturing Origin | United States |
| Media Type | Lowenstein-Jensen Medium (LJ) |
| Product Type | Microbiology Media |
| Quantity | 100/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction