Learn More
Invitrogen™ LOX-1 Monoclonal Antibody (01)

Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human OLR1/LOX1.
Specifications
Specifications
| Antigen | LOX-1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 01 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | OLR1 |
| Gene Accession No. | P78380 |
| Gene Alias | CLEC8A; C-type lectin domain family 8 member A; hLOX 1; hLOX1; hLOX-1; lectin-like oxidized LDL receptor; lectin-like oxidized LDL receptor 1; lectin-like oxidized low-density lipoprotein receptor; Lectin-like oxidized low-density lipoprotein receptor-1; lectin-like oxLDL receptor 1; lectin-type oxidized LDL receptor 1; LOX 1; LOX1; LOX-1; LOXIN; Oldlr1; Oldr1; OLR1; ox LDL receptor 1; oxidised low density lipoprotein (lectin-like) receptor 1; oxidized low density lipoprotein (lectin-like) receptor 1; oxidized low density lipoprotein receptor 1; oxidized low-density lipoprotein receptor 1; Oxidized low-density lipoprotein receptor 1, soluble form; ox-LDL receptor 1; Scare1; scavenger receptor class E, member 1; sLOX 1; SLOX1; soluble LOX 1; SR-EI |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.