Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LOX-1/OLR1 Antibody (014), Alexa Fluor™ 532, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290950AF532
Description
LOX-1/OLR1 Monoclonal specifically detects LOX-1/OLR1 in Cynomolgus Monkey samples. It is validated for ELISA.Specifications
LOX-1/OLR1 | |
Monoclonal | |
Alexa Fluor 532 | |
CLEC8ASLOX1, C-type lectin domain family 8 member A, hLOX-1, Lectin-like oxidized LDL receptor 1, Lectin-like oxLDL receptor 1, Lectin-type oxidized LDL receptor 1, LOX-1, LOX1ox LDL receptor 1, LOXIN, oxidised low density lipoprotein (lectin-like) receptor 1, oxidized low density lipoprotein (lectin-like) receptor 1, oxidized low-density lipoprotein receptor 1, oxidized low-density lipoprotein receptor 1, soluble form, Ox-LDL receptor 1, SCARE1, scavenger receptor class E, member 1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Cynomolgus LOX-1/OLR1 (G7PJT2; Ser61-Gln273). | |
0.1 mL | |
Cardiovascular Biology, metabolism | |
4973 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
14 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Cynomolgus Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction