Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LOX 1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984462
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4462 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4462 Supplier Bioss Supplier No. BS2044R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LOX-1 (lectin-like oxidized LDL receptor -1) is a 40-50 kDa type II membrane glycoprotein class E scavenger receptor expressed by endothelial cells, macrophages, activated platelets and smooth muscle cells. LOX-1 binds to a wide range of ligands, including oxidized low-density lipoprotein (oxLDL), hypochlorite modified high-density lipoprotein (HDL), aged /apoptotic cells, activated platelets and bacteria, reflecting its versatile physiological functions.

Specifications

Antigen LOX 1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene OLR1
Gene Accession No. P78380
Gene Alias CLEC8A; C-type lectin domain family 8 member A; hLOX 1; hLOX1; hLOX-1; Lectin-like oxidized LDL receptor 1; lectin-like oxidized low-density lipoprotein receptor; Lectin-like oxidized low-density lipoprotein receptor-1; lectin-like oxLDL receptor 1; Lectin-type oxidized LDL receptor 1; LOX 1; Lox1; LOX-1; LOXIN; Oldlr1; Oldr1; Olr1; ox LDL receptor 1; oxidised low density lipoprotein (lectin-like) receptor 1; oxidized low density lipoprotein (lectin-like) receptor 1; oxidized low density lipoprotein receptor 1; Oxidized low-density lipoprotein receptor 1; Oxidized low-density lipoprotein receptor 1, soluble form; Ox-LDL receptor 1; SCARE1; scavenger receptor class E, member 1; sLOX 1; SLOX1; soluble LOX 1; SR-EI
Gene Symbols OLR1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from Rabbit LOX-1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4973
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.