Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LOX Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002530AF488
Description
LOX Polyclonal specifically detects LOX in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
LOX | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
P28300 | |
LOX | |
A cocktail of two synthetic peptides; one made to a region of the human LOX protein within residues 300-350 (NB100-2528) and one within residues 200-300 (NB100-2527). [Swiss-Prot P28300] | |
0.05 mL | |
Cancer, Cytoskeleton Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Signal Transduction | |
4015 | |
Human, Mouse, Rat, Pig, Bovine | |
Precipitation |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
lysyl oxidaseEC 1.4.3.13, MGC105112, protein-lysine 6-oxidase | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, mouse, rat, cow and pig. 92% sequence identity with chicken, Zebrafish, and Xenopus proteins. | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction