Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LOX Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 15 publications
$243.50 - $390.00
Specifications
Antigen | LOX |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:500-1:1000, Simple Western 1:100, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:200 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB1002530SS
![]() |
Novus Biologicals
NB1002530SS |
0.025 mL |
Each for $243.50
|
|
|||||
NB1002530
![]() |
Novus Biologicals
NB1002530 |
0.05 mL |
Each for $390.00
|
|
|||||
Description
LOX Polyclonal specifically detects LOX in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
LOX | |
Western Blot 1:500-1:1000, Simple Western 1:100, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:200 | |
Polyclonal | |
Precipitation | |
RUO | |
Human, Mouse, Rat, Pig, Bovine | |
P28300 | |
4015 | |
A cocktail of two synthetic peptides; one made to a region of the human LOX protein within residues 300-350 (NB100-2528) and one within residues 200-300 (NB100-2527). [Swiss-Prot P28300] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
1.0 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cancer, Cytoskeleton Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Signal Transduction | |
PBS with 0.02% Sodium Azide | |
lysyl oxidaseEC 1.4.3.13, MGC105112, protein-lysine 6-oxidase | |
LOX | |
IgG | |
Immunogen affinity purified | |
Human, mouse, rat, cow and pig. 92% sequence identity with chicken, Zebrafish, and Xenopus proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title