Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LOX Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 27 publications

$269.00 - $548.00

Specifications

Antigen LOX
Concentration 1.0 mg/ml
Dilution Western Blot 0.5-4ug/ml, Simple Western 1:25, Immunohistochemistry 1:100-1:250, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100-1:250
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB1002527SS
SDP
View Documents
Novus Biologicals
NB1002527SS
0.025 mL
Each for $269.00
Only null left
Add to Cart
 
NB1002527
SDP
View Documents
Novus Biologicals
NB1002527
0.1 mL
Each for $548.00
Only null left
Add to Cart
 
Description

Description

LOX Polyclonal specifically detects LOX in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications

Specifications

LOX
Western Blot 0.5-4ug/ml, Simple Western 1:25, Immunohistochemistry 1:100-1:250, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100-1:250
Polyclonal
Rabbit
Cancer, Cytoskeleton Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Signal Transduction
Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide
lysyl oxidaseEC 1.4.3.13, MGC105112, protein-lysine 6-oxidase
LOX
IgG
Affinity Purified
Human and mouse. Predicted to react with rat and pig based on 100% sequence homology. Immunogen sequence has 92% homology to cow and chicken and 85% homology to Xenopus.
1.0 mg/ml
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse, Rat
P28300
4015
A synthetic peptide made to an internal region of the human LOX protein (within residues 200-300). [Swiss-Prot P28300].
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.