Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LOX Recombinant Rabbit Monoclonal Antibody (9K10Z3)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA535123
Description
Immunogen sequence: GHKASFCLED TSCDYGYHRR FACTAHTQGL SPGCYDTYGA DIDCQWIDIT DVKPGNYILK VSVNPSYLVP ESDYTNNVVR CDIRYTGHHA YASGCTISPY.
Lysyl oxidase (LOX), a copper-containing amine oxidase, belongs to a heterogeneous family of enzymes that oxidize primary amine substrates to reactive aldehydes. It plays a vital role in the formation and repair of the extra cellular matrix. As well, LOX is a multifunctional enzyme having diverse biological functions such as developmental regulation, tumor suppression, cell motility and cellular senescence. The secreted form of LOX is responsible for the invasive properties of hypoxic human cancer cells. Thus, it is essential for hypoxia-induced metastasis and is a good therapeutic target for preventing and treating metastases.
Specifications
LOX | |
Recombinant Monoclonal | |
0.5 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P16636, P28300, P28301 | |
LOX | |
A synthetic peptide corresponding to a sequence within amino acids 318-417 of human LOX (P28300). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
9K10Z3 | |
Unconjugated | |
LOX | |
AI893619; cross-linking enzyme; H-rev142; LOX; Lysyl oxidase; Lysyl oxidase (an H-rev gene with its expression down-regulated in HRAS-transformed rat 208F fibroblasts); MGC105112; protein-lysine 6-oxidase; Protein-lysine 6-oxidase, long form; Protein-lysine 6-oxidase, short form; Ras excision protein; ras recision; rrg; Rrg1; TSC-160 | |
Rabbit | |
Affinity chromatography | |
RUO | |
16948, 24914, 4015 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction