Learn More
Invitrogen™ LOX Polyclonal Antibody

Description
PA1-16953 is specific for the ~58 kDa LOX protein. This is a highly glycosylated protein and therefore, other bands may be seen at ~32 kDa (non-glycosylated form) 50 kDa and 58 kDa (glycosylated forms). Suggested positive control: human and mouse kidney lysate, antigen standard for LOX (transient overexpression lysate), human kidney protein, mouse kidney protein.
Specifications
Specifications
| Antigen | LOX |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | LOX |
| Gene Accession No. | P16636, P28300, P28301, P45845 |
| Gene Alias | AI893619; cross-linking enzyme; H-rev142; LOX; Lysyl oxidase; Lysyl oxidase (an H-rev gene with its expression down-regulated in HRAS-transformed rat 208F fibroblasts); MGC105112; protein-lysine 6-oxidase; Protein-lysine 6-oxidase, long form; Protein-lysine 6-oxidase, short form; Ras excision protein; ras recision; rrg; Rrg1; TSC-160 |
| Gene Symbols | LOX |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.