Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LOX Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01950184
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-950-184 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-950-184 Supplier Invitrogen™ Supplier No. PA116955
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is 92% homologous to chicken, zebrafish, and xenopus.

Lysyl oxidase (LOX), a copper-containing amine oxidase, belongs to a heterogeneous family of enzymes that oxidize primary amine substrates to reactive aldehydes. It plays a vital role in the formation and repair of the extra cellular matrix. As well, LOX is a multifunctional enzyme having diverse biological functions such as developmental regulation, tumor suppression, cell motility and cellular senescence. The secreted form of LOX is responsible for the invasive properties of hypoxic human cancer cells. Thus, it is essential for hypoxia-induced metastasis and is a good therapeutic target for preventing and treating metastases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LOX
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene LOX
Gene Accession No. P16636, P28300, P28301, P33072, P45845
Gene Alias AI893619; cross-linking enzyme; H-rev142; LOX; Lysyl oxidase; Lysyl oxidase (an H-rev gene with its expression down-regulated in HRAS-transformed rat 208F fibroblasts); MGC105112; protein-lysine 6-oxidase; Protein-lysine 6-oxidase, long form; Protein-lysine 6-oxidase, short form; Ras excision protein; ras recision; rrg; Rrg1; TSC-160
Gene Symbols LOX
Host Species Rabbit
Immunogen Cocktail of two synthetic peptides between amino acids 300-350 and amino acids 200-300 of the human LOX protein.
Purification Method Antigen affinity chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100525278, 16948, 24914, 280841, 4015
Target Species Human, Mouse, Rat, Bovine, Pig
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.