Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LOX propeptide Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP130327MFV610
Description
LOX propeptide Polyclonal antibody specifically detects LOX propeptide in Human, Rat samples. It is validated for Flow CytometrySpecifications
LOX propeptide | |
Polyclonal | |
Flow Cytometry | |
lysyl oxidaseEC 1.4.3.13, MGC105112, protein-lysine 6-oxidase | |
Synthetic peptide made to an internal portion of human LOX propeptide (within residues 118-168). [Swiss-Prot# P28300] | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Signal Transduction | |
4015 | |
Store at 4C in the dark. | |
IgG |
Flow Cytometry | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction