Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LOX Rabbit anti-Human, Mouse, Rat, Clone: 9K10Z3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | LOX |
---|---|
Clone | 9K10Z3 |
Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunoprecipitation, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
LOX Monoclonal antibody specifically detects LOX in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
LOX | |
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunoprecipitation, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
lysyl oxidaseEC 1.4.3.13, MGC105112, protein-lysine 6-oxidase | |
A synthetic peptide corresponding to a sequence within amino acids 318-417 of human LOX (P28300). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
9K10Z3 | |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
4015 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title