Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LOXHD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144260
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144260 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144260 Supplier Invitrogen™ Supplier No. PA5144260
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LOXHD1 Polyclonal Antibody for ICC/IF, IHC (P), ELISA

Involved in hearing. Required for normal function of hair cells in the inner ear.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LOXHD1
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene LOXHD1
Gene Accession No. Q8IVV2
Gene Alias Lipoxygenase homology domain-containing protein 1; LOXHD1
Gene Symbols LOXHD1
Host Species Rabbit
Immunogen Recombinant Human Lipoxygenase homology domain-containing protein 1 protein (1101-1366AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 125336
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.