Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LOXL1 Monoclonal Antibody (C1)
GREENER_CHOICE

Catalog No. PIMA541635
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541635 200 μL
PIMA543676 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA541635 Supplier Invitrogen™ Supplier No. MA541635
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

LOXL1 Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes a member of the lysyl oxidase gene family. The prototypic member of the family is essential to the biogenesis of connective tissue, encoding an extracellular copper-dependent amine oxidase that catalyses the first step in the formation of crosslinks in collagens and elastin. A highly conserved amino acid sequence at the C-terminus end appears to be sufficient for amine oxidase activity, suggesting that each family member may retain this function. The N-terminus is poorly conserved and may impart additional roles in developmental regulation, senescence, tumor suppression, cell growth control, and chemotaxis to each member of the family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LOXL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone C1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Loxl1
Gene Accession No. Q08397
Gene Alias cross-linking enzyme; LOL; Lox2; LOXL; LOXL1; Lysyl oxidase 2; lysyl oxidase homolog 1; lysyl oxidase like 1; lysyl oxidase-like 1; lysyl oxidase-like protein 1
Gene Symbols Loxl1
Host Species Mouse
Immunogen Recombinant protein Lysyl Oxidase Like Protein 1. The antigen corresponds to amino acid range 369-574 of the target protein.
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4016
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.