Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LOXL2 Monoclonal Antibody (15)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA530651
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human LOXL2/Lysyl oxidase-like 2.
This gene encodes a member of the lysyl oxidase gene family. The prototypic member of the family is essential to the biogenesis of connective tissue, encoding an extracellular copper-dependent amine oxidase that catalyses the first step in the formation of crosslinks in collagens and elastin. A highly conserved amino acid sequence at the C-terminus end appears to be sufficient for amine oxidase activity, suggesting that each family member may retain this function. The N-terminus is poorly conserved and may impart additional roles in developmental regulation, senescence, tumor suppression, cell growth control, and chemotaxis to each member of the family.
Specifications
LOXL2 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q9Y4K0 | |
LOXL2 | |
Recombinant Human LOXL2/Lysyl oxidase-like 2 Protein (Met1-Gln774). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
15 | |
Unconjugated | |
LOXL2 | |
1110004B06Rik; 4930526G11Rik; 9430067E15Rik; LOR2; Loxl2; Lysyl oxidase homolog 2; lysyl oxidase homolog 2; LOW QUALITY PROTEIN: lysyl oxidase homolog 2; lysyl oxidase like 2; lysyl oxidase related 2; lysyl oxidase-like 2; lysyl oxidase-like 2 delta e13; lysyl oxidase-like 2 protein; lysyl oxidase-like protein 2; Lysyl oxidase-related protein 2; lysyl oxidase-related protein WS9-14; WS9-14 | |
Mouse | |
Protein A | |
RUO | |
4017 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction