Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LOXL2 Monoclonal Antibody (262418)
GREENER_CHOICE

Catalog No. PIMA523963
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA523963 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA523963 Supplier Invitrogen™ Supplier No. MA523963
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute at 0.5 mg/mL in sterile PBS.

This gene encodes a member of the lysyl oxidase gene family. The prototypic member of the family is essential to the biogenesis of connective tissue, encoding an extracellular copper-dependent amine oxidase that catalyses the first step in the formation of crosslinks in collagens and elastin. A highly conserved amino acid sequence at the C-terminus end appears to be sufficient for amine oxidase activity, suggesting that each family member may retain this function. The N-terminus is poorly conserved and may impart additional roles in developmental regulation, senescence, tumor suppression, cell growth control, and chemotaxis to each member of the family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LOXL2
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 262418
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene LOXL2
Gene Accession No. Q9Y4K0
Gene Alias 1110004B06Rik; 4930526G11Rik; 9430067E15Rik; LOR2; Loxl2; Lysyl oxidase homolog 2; lysyl oxidase homolog 2; LOW QUALITY PROTEIN: lysyl oxidase homolog 2; lysyl oxidase like 2; lysyl oxidase related 2; lysyl oxidase-like 2; lysyl oxidase-like 2 delta e13; lysyl oxidase-like 2 protein; lysyl oxidase-like protein 2; Lysyl oxidase-related protein 2; lysyl oxidase-related protein WS9-14; WS9-14
Gene Symbols LOXL2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Lysyl Oxidase Homolog 2/LOXL2 Gln26-Gln744 (predicted).
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4017
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.