Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LOXL2 Monoclonal Antibody (OTI4H3), TrueMAB™, OriGene
SDP

Catalog No. 501681553
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-1553 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-1553 Supplier OriGene Supplier No. TA807444
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lysyl Oxidase (LO) is an extracellular, copper-dependent enzyme that initiates the cross-linking of collagens and elastin by catalyzing the oxidative deamination of peptidyl lysine to alpha-aminoadipic-delta-semialdehyde. Members of LO family have diverse functions which include tumor suppression and cell adhesion and senescence. LOXL2, a 87KDa protein, is a novel member of this family. It contains 3 potential N-linked glycosylation sites, 4 scavenger receptor cysteine-rich (SRCR) domains and highly conserved copper-binding and Lysyl-tyrosylquinone cofactor sites. It is a component of extracellular matrix that might be involved in cell adhesion and senescence. Role of LOXL2 has been implicated in maligt transformation.

Specifications

Antigen LOXL2
Applications Western Blot
Classification Monoclonal
Clone OTI4H3
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene LOXL2
Gene Accession No. Q9Y4K0
Gene Alias LOR, LOR2, WS9-14
Gene Symbols LOXL2
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 152-450 of human LOXL2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4017
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.