Learn More
LOXL2 Monoclonal Antibody (OTI8G5)

Description
LOXL2 Monoclonal Antibody for IHC (P)
Specifications
Specifications
| Antigen | LOXL2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | OTI8G5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | LOXL2 |
| Gene Accession No. | Q9Y4K0 |
| Gene Alias | 1110004B06Rik; 4930526G11Rik; 9430067E15Rik; LOR2; Loxl2; Lysyl oxidase homolog 2; lysyl oxidase homolog 2; LOW QUALITY PROTEIN: lysyl oxidase homolog 2; lysyl oxidase like 2; lysyl oxidase related 2; lysyl oxidase-like 2; lysyl oxidase-like 2 delta e13; lysyl oxidase-like 2 protein; lysyl oxidase-like protein 2; Lysyl oxidase-related protein 2; lysyl oxidase-related protein WS9-14; WS9-14 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.