Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LOXL4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550390
Description
The antibody detects endogenous levels of total LOXL4 protein.
This gene encodes a member of the lysyl oxidase gene family. The prototypic member of the family is essential to the biogenesis of connective tissue, encoding an extracellular copper-dependent amine oxidase that catalyses the first step in the formation of crosslinks in collagens and elastin. A highly conserved amino acid sequence at the C-terminus end appears to be sufficient for amine oxidase activity, suggesting that each family member may retain this function. The N-terminus is poorly conserved and may impart additional roles in developmental regulation, senescence, tumor suppression, cell growth control, and chemotaxis to each member of the family.
Specifications
LOXL4 | |
Polyclonal | |
Unconjugated | |
LOXL4 | |
4833426I20Rik; FLJ21889; Loxc; LOXL4; Lysyl oxidase homolog 4; lysyl oxidase like 4; lysyl oxidase related C; lysyl oxidase-like 4; lysyl oxidase-like 4 pseudogene; lysyl oxidase-like protein 4; Lysyl oxidase-related protein C | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84171 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q96JB6 | |
LOXL4 | |
Full length fusion protein of Human LOXL4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction