Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LOXL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558036
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558036 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558036 Supplier Invitrogen™ Supplier No. PA558036
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RRHRGYLSET VSNALGPQGR RLEEVRLKPI LASAKQHSPV TEGAVEVKYE GHWRQVCDQG WTMN Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 91%.

This gene encodes a member of the lysyl oxidase gene family. The prototypic member of the family is essential to the biogenesis of connective tissue, encoding an extracellular copper-dependent amine oxidase that catalyses the first step in the formation of crosslinks in collagens and elastin. A highly conserved amino acid sequence at the C-terminus end appears to be sufficient for amine oxidase activity, suggesting that each family member may retain this function. The N-terminus is poorly conserved and may impart additional roles in developmental regulation, senescence, tumor suppression, cell growth control, and chemotaxis to each member of the family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LOXL4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LOXL4
Gene Accession No. Q96JB6
Gene Alias 4833426I20Rik; FLJ21889; Loxc; LOXL4; Lysyl oxidase homolog 4; lysyl oxidase like 4; lysyl oxidase related C; lysyl oxidase-like 4; lysyl oxidase-like 4 pseudogene; lysyl oxidase-like protein 4; Lysyl oxidase-related protein C
Gene Symbols LOXL4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LOXL4. Recombinant protein control fragment (Product #RP-97095).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84171
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.