Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LOXL4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | LOXL4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
LOXL4 Polyclonal specifically detects LOXL4 in Human samples. It is validated for Western Blot.Specifications
LOXL4 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
EC 1.4.3, EC 1.4.3.-, FLJ21889, LOXClysyl oxidase related C, lysyl oxidase homolog 4, lysyl oxidase-like 4, Lysyl oxidase-like protein 4, Lysyl oxidase-related protein C | |
Recombinant fusion protein containing a sequence corresponding to amino acids 25-280 of human LOXL4 (NP_115587.6). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism | |
PBS with 50% glycerol, pH7.3. | |
84171 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title